Assay Details
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172410.2 | NP_765998.1 | 21 - 22 | 2321 | 68 | |
GenBank mRNA | AK010450.1 | - | 20 - 21 | 2272 | 68 | |
AK028864.1 | - | 21 - 22 | 2322 | 68 | ||
AK045380.1 | - | 2180 | 68 | |||
AK129055.1 | - | 19 - 20 | 2665 | 68 | ||
AK167105.1 | - | 20 - 21 | 2272 | 68 | ||
AK169194.1 | - | 18 - 19 | 2042 | 68 | ||
AK169693.1 | - | 20 - 21 | 2291 | 68 | ||
BC023140.1 | - | 20 - 21 | 2267 | 3991335 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
