Assay Details
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172443.3 | NP_766031.2 | 1 - 2 | 107 | 131 | |
GenBank mRNA | AK047555.1 | - | 1 - 2 | 108 | 131 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_172443.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
