Assay Details
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172443.3 | NP_766031.2 | 4 - 5 | 1101 | 102 | |
GenBank mRNA | AK047555.1 | - | 4 - 5 | 1102 | 102 | |
BC057634.1 | - | 4 - 5 | 1063 | 2866041 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
