Assay Details
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009188.4 | NP_033214.2 | 7 - 8 | 954 | 70 | |
GenBank mRNA | AK159076.1 | - | 7 - 8 | 937 | 70 | |
AK173005.1 | - | 6 - 7 | 2904 | 70 | ||
BC129859.1 | - | 7 - 8 | 926 | 70 | ||
L38622.2 | - | 7 - 8 | 926 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
