Assay Details
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009188.4 | NP_033214.2 | 11 - 12 | 1638 | 87 | |
GenBank mRNA | AK087970.1 | - | 4 - 5 | 735 | 87 | |
AK173005.1 | - | 10 - 11 | 3588 | 87 | ||
BC129859.1 | - | 11 - 12 | 1610 | 87 | ||
L38622.2 | - | 11 - 12 | 1610 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
