Assay Details
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009188.4 | NP_033214.2 | 14 - 15 | 2540 | 66 | |
GenBank mRNA | AK173005.1 | - | 13 - 14 | 4490 | 66 | |
BC129859.1 | - | 14 - 15 | 2512 | 66 | ||
L38622.2 | - | 14 - 15 | 2512 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
