Assay Details
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009188.4 | NP_033214.2 | 16 - 17 | 2807 | 72 | |
GenBank mRNA | AK087970.1 | - | 9 - 10 | 1904 | 72 | |
AK159076.1 | - | 16 - 17 | 2790 | 72 | ||
AK173005.1 | - | 15 - 16 | 4757 | 72 | ||
BC129859.1 | - | 16 - 17 | 2779 | 72 | ||
L38622.2 | - | 16 - 17 | 2779 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
