Assay Details
Gene Symbol: Bap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027088.2 | NP_081364.1 | 7 - 8 | 706 | 100 | |
GenBank mRNA | AB047820.1 | - | 7 - 8 | 607 | 100 | |
AK079374.1 | - | 7 - 8 | 722 | 100 | ||
AK129108.1 | - | 7 - 8 | 614 | 100 | ||
AK170576.1 | - | 7 - 8 | 707 | 100 | ||
AK171561.1 | - | 7 - 8 | 708 | 100 | ||
AK185783.1 | - | 1 - 2 | 91 | 100 | ||
AK189418.1 | - | 1 - 2 | 92 | 100 | ||
AK193408.1 | - | 1 - 2 | 91 | 100 | ||
BC042489.1 | - | 7 - 8 | 657 | 100 | ||
BC050901.1 | - | 7 - 8 | 669 | 6402443 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
