Assay Details
Gene Symbol: Tdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028354.4 | NP_082630.2 | 3 - 4 | 644 | 61 | |
GenBank mRNA | AK014855.1 | - | 3 - 4 | 624 | 61 | |
AK033868.1 | - | 3 - 4 | 642 | 61 | ||
BC065162.1 | - | 2 - 3 | 614 | 6402921 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
