Assay Details
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC027320.1 | - | 6 - 7 | 1149 | 67 | |
BC037012.1 | - | 6 - 7 | 1394 | 5342428 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
