Assay Details
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172473.3 | NP_766061.2 | 9 - 10 | 1060 | 89 | |
GenBank mRNA | AK042879.1 | - | 9 - 10 | 1001 | 89 | |
AK156958.1 | - | 9 - 10 | 1002 | 89 | ||
BC120695.1 | - | 9 - 10 | 957 | 40129618 | 89 | |
BC120697.1 | - | 9 - 10 | 957 | 40129620 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
