Assay Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111267.2 | NP_001104737.1 | 3 - 4 | 516 | 125 | |
NM_172495.5 | NP_766083.3 | 13 - 14 | 2867 | 125 | ||
GenBank mRNA | AK033503.1 | - | 3 - 4 | 430 | 125 | |
AK042369.1 | - | 3 - 4 | 486 | 125 | ||
AK128935.1 | - | 12 - 13 | 2690 | 125 | ||
AK165098.1 | - | 3 - 4 | 485 | 125 | ||
AK171679.1 | - | 3 - 4 | 434 | 125 | ||
BC042791.1 | - | 947 | 125 | |||
BC049997.1 | - | 3 - 4 | 466 | 125 | ||
BC058572.1 | - | 3 - 4 | 438 | 125 | ||
BC076623.1 | - | 11 - 12 | 2473 | 125 | ||
BC080670.1 | - | 440 | 125 | |||
BC141150.1 | - | 12 - 13 | 2568 | 9055762 | 125 | |
BC145459.1 | - | 11 - 12 | 2453 | 125 | ||
EF602028.1 | - | 3 - 4 | 516 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
