Assay Details
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172560.3 | NP_766148.2 | 6 - 7 | 1720 | 79 | |
GenBank mRNA | AK036710.1 | - | 5 - 6 | 1540 | 79 | |
BC058512.1 | - | 6 - 7 | 553 | 6817014 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
