Assay Details
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167680.1 | NP_001161152.1 | 8 - 9 | 1219 | 54 | |
NM_172572.3 | NP_766160.2 | 8 - 9 | 1433 | 54 | ||
GenBank mRNA | AK028872.1 | - | 8 - 9 | 1434 | 54 | |
AK042365.1 | - | 7 - 8 | 1886 | 54 | ||
AK154639.1 | - | 8 - 9 | 1199 | 54 | ||
AK171085.1 | - | 8 - 9 | 1215 | 54 | ||
AY167043.1 | - | 8 - 9 | 1219 | 54 | ||
BC031398.1 | - | 2 - 3 | 252 | 54 | ||
BC052182.1 | - | 8 - 9 | 1167 | 6335022 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
