Assay Details
Gene Symbol: Pogz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165948.1 | NP_001159420.1 | 11 - 12 | 1915 | 93 | |
NM_172683.3 | NP_766271.2 | 11 - 12 | 1942 | 93 | ||
GenBank mRNA | AK035255.1 | - | 11 - 12 | 1797 | 93 | |
AK122288.1 | - | 1 - 2 | 99 | 93 | ||
AK161889.1 | - | 11 - 12 | 1770 | 93 | ||
BC032176.1 | - | 2 - 3 | 233 | 93 | ||
BC065134.1 | - | 11 - 12 | 1931 | 93 | ||
BC066108.1 | - | 11 - 12 | 1931 | 93 | ||
BC085188.1 | - | 11 - 12 | 1775 | 30682742 | 93 | |
BC096492.1 | - | 10 - 11 | 1785 | 30745658 | 93 | |
BC139432.1 | - | 11 - 12 | 1797 | 8862454 | 93 | |
BC139433.1 | - | 11 - 12 | 1797 | 8862455 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
