Assay Details
Gene Symbol: Pogz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165948.1 | NP_001159420.1 | 13 - 14 | 2202 | 90 | |
NM_172683.3 | NP_766271.2 | 13 - 14 | 2229 | 90 | ||
GenBank mRNA | AK030880.1 | - | 1 - 2 | 125 | 90 | |
AK035255.1 | - | 13 - 14 | 2084 | 90 | ||
AK122288.1 | - | 3 - 4 | 386 | 90 | ||
AK161889.1 | - | 13 - 14 | 2057 | 90 | ||
BC032176.1 | - | 4 - 5 | 520 | 90 | ||
BC065134.1 | - | 13 - 14 | 2218 | 90 | ||
BC066108.1 | - | 13 - 14 | 2218 | 90 | ||
BC085188.1 | - | 13 - 14 | 2062 | 30682742 | 90 | |
BC096492.1 | - | 12 - 13 | 2072 | 30745658 | 90 | |
BC139432.1 | - | 13 - 14 | 2084 | 8862454 | 90 | |
BC139433.1 | - | 13 - 14 | 2084 | 8862455 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
