Assay Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009316.1 | NP_033342.1 | 7 - 8 | 893 | 94 | |
NM_172688.3 | NP_766276.1 | 7 - 8 | 893 | 94 | ||
GenBank mRNA | AK046565.1 | - | 7 - 8 | 859 | 94 | |
AK053911.1 | - | 7 - 8 | 879 | 94 | ||
AK137046.1 | - | 7 - 8 | 894 | 94 | ||
AK159277.1 | - | 7 - 8 | 857 | 94 | ||
BC006665.1 | - | 7 - 8 | 884 | 3499247 | 94 | |
D76446.1 | - | 7 - 8 | 893 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
