Assay Details
Gene Symbol: Sel1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172710.3 | NP_766298.2 | 18 - 19 | 2923 | 98 | |
GenBank mRNA | AK030487.1 | - | 18 - 19 | 2788 | 98 | |
AK122362.1 | - | 18 - 19 | 2923 | 98 | ||
BC026399.1 | - | 1 - 2 | 76 | 98 | ||
BC026655.1 | - | 6 - 7 | 646 | 98 | ||
BC094607.1 | - | 17 - 18 | 2468 | 6837048 | 98 | |
BC117915.1 | - | 17 - 18 | 2424 | 40097358 | 98 | |
BC117916.1 | - | 17 - 18 | 2426 | 40097359 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
