Assay Details
Gene Symbol: Chfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289577.1 | NP_001276506.1 | 1 - 2 | 101 | 73 | |
NM_001289578.1 | NP_001276507.1 | 1 - 2 | 101 | 73 | ||
NM_001289579.1 | NP_001276508.1 | 1 - 2 | 101 | 73 | ||
NM_001289580.1 | NP_001276509.1 | 1 - 2 | 101 | 73 | ||
NM_172717.4 | NP_766305.2 | 1 - 2 | 101 | 73 | ||
GenBank mRNA | AK039281.1 | - | 1 - 2 | 90 | 73 | |
AK048930.1 | - | 1 - 2 | 81 | 73 | ||
AK052473.1 | - | 1 - 2 | 56 | 73 | ||
AK077629.1 | - | 1 - 2 | 73 | 73 | ||
AK133154.1 | - | 1 - 2 | 83 | 73 | ||
AK147892.1 | - | 1 - 2 | 76 | 73 | ||
AK148436.1 | - | 1 - 2 | 72 | 73 | ||
AK154038.1 | - | 1 - 2 | 83 | 73 | ||
AK155295.1 | - | 1 - 2 | 73 | 73 | ||
AK155525.1 | - | 1 - 2 | 73 | 73 | ||
AK169182.1 | - | 1 - 2 | 54 | 73 | ||
AK169193.1 | - | 1 - 2 | 56 | 73 | ||
BC049792.1 | - | 1 - 2 | 55 | 6510480 | 73 | |
BY142662.1 | - | 1 - 2 | 91 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK052473.1, AK077629.1, AK133154.1, AK147892.1, AK154038.1, AK155295.1, AK155525.1, AK169182.1, AK169193.1, BC049792.1, NM_001289577.1, NM_001289578.1, NM_001289579.1, NM_001289580.1, NM_172717.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
