Assay Details
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172721.2 | NP_766309.2 | 6 - 7 | 1106 | 69 | |
GenBank mRNA | AK031815.1 | - | 6 - 7 | 1100 | 69 | |
AK034447.1 | - | 6 - 7 | 1034 | 69 | ||
AK047695.1 | - | 6 - 7 | 1074 | 69 | ||
AK047756.1 | - | 6 - 7 | 1048 | 69 | ||
AK053794.1 | - | 6 - 7 | 1107 | 69 | ||
AK134291.1 | - | 6 - 7 | 1113 | 69 | ||
AK214176.1 | - | 1 - 2 | 67 | 69 | ||
BC009095.1 | - | 6 - 7 | 1059 | 3158218 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
