Assay Details
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172722.3 | NP_766310.2 | 3 - 4 | 313 | 68 | |
GenBank mRNA | AK049325.1 | - | 3 - 4 | 314 | 68 | |
AK052060.1 | - | 3 - 4 | 301 | 68 | ||
AK149758.1 | - | 3 - 4 | 314 | 68 | ||
AK165175.1 | - | 3 - 4 | 304 | 68 | ||
AK167405.1 | - | 3 - 4 | 331 | 68 | ||
AK167491.1 | - | 3 - 4 | 331 | 68 | ||
BC028868.1 | - | 1 - 2 | 93 | 68 | ||
BC037595.1 | - | 3 - 4 | 294 | 68 | ||
BC055480.1 | - | 2 - 3 | 215 | 68 | ||
BC082314.1 | - | 3 - 4 | 299 | 68 | ||
BC106182.1 | - | 3 - 4 | 288 | 68 | ||
BC132172.1 | - | 3 - 4 | 311 | 40130449 | 68 | |
BC138093.1 | - | 3 - 4 | 311 | 8861112 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
