Assay Details
Gene Symbol: Ttc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172770.3 | NP_766358.1 | 11 - 12 | 988 | 64 | |
GenBank mRNA | AK054350.1 | - | 11 - 12 | 989 | 64 | |
BC056616.1 | - | 10 - 11 | 916 | 4974294 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
