Assay Details
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172780.3 | NP_766368.2 | 8 - 9 | 1175 | 87 | |
GenBank mRNA | AK032326.1 | - | 8 - 9 | 1156 | 87 | |
AK122232.1 | - | 8 - 9 | 1146 | 87 | ||
BC130221.1 | - | 8 - 9 | 1160 | 40141414 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
