Assay Details
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172787.2 | NP_766375.1 | 15 - 16 | 1607 | 58 | |
GenBank mRNA | AK030412.1 | - | 2 - 3 | 205 | 58 | |
AK045667.1 | - | 15 - 16 | 1608 | 58 | ||
AK049597.1 | - | 15 - 16 | 1642 | 58 | ||
AK129443.1 | - | 11 - 12 | 1152 | 58 | ||
BC082309.1 | - | 14 - 15 | 1477 | 30356672 | 58 | |
BC085192.1 | - | 14 - 15 | 1712 | 30694487 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
