Assay Details
Gene Symbol: Gucy1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204340.1 | NP_001191269.1 | 8 - 9 | 1032 | 68 | |
NM_172810.3 | NP_766398.1 | 8 - 9 | 1077 | 68 | ||
GenBank mRNA | AF038500.1 | - | 2 - 3 | 231 | 68 | |
AK047129.1 | - | 8 - 9 | 1078 | 68 | ||
AY236513.1 | - | 6 - 7 | 751 | 68 | ||
BC137868.1 | - | 8 - 9 | 1029 | 8860887 | 68 | |
BC144988.1 | - | 8 - 9 | 1026 | 9053536 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
