Assay Details
Gene Symbol: Slc4a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271544.1 | NP_001258473.1 | 15 - 16 | 2187 | 65 | |
NM_001271545.1 | NP_001258474.1 | 15 - 16 | 2187 | 65 | ||
NM_001271546.1 | NP_001258475.1 | 15 - 16 | 2187 | 65 | ||
NM_001271547.1 | NP_001258476.1 | 15 - 16 | 2187 | 65 | ||
NM_172830.2 | NP_766418.1 | 14 - 15 | 1971 | 65 | ||
GenBank mRNA | AK085269.1 | - | 14 - 15 | 1971 | 65 | |
AK085424.1 | - | 15 - 16 | 2192 | 65 | ||
AK128930.1 | - | 14 - 15 | 2327 | 65 | ||
AK142545.1 | - | 15 - 16 | 2189 | 65 | ||
BC113179.1 | - | 15 - 16 | 2182 | 40060674 | 65 | |
BC113785.1 | - | 15 - 16 | 2181 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
