Assay Details
Gene Symbol: Slc13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172892.3 | NP_766480.1 | 14 - 15 | 2277 | 63 | |
GenBank mRNA | AK036354.1 | - | 14 - 15 | 2279 | 63 | |
AK038937.1 | - | 14 - 15 | 2282 | 63 | ||
BC089161.1 | - | 14 - 15 | 2266 | 30440173 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
