Assay Details
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172926.3 | NP_766514.2 | 9 - 10 | 1006 | 85 | |
GenBank mRNA | AK046839.1 | - | 9 - 10 | 948 | 85 | |
AK049371.1 | - | 9 - 10 | 985 | 85 | ||
AK082389.1 | - | 8 - 9 | 900 | 85 | ||
BC080799.1 | - | 9 - 10 | 991 | 4948538 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
