Assay Details
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172964.4 | NP_766552.3 | 12 - 13 | 1645 | 70 | |
GenBank mRNA | AK087523.1 | - | 12 - 13 | 1658 | 70 | |
AK133720.1 | - | 12 - 13 | 1484 | 70 | ||
AK141901.1 | - | 12 - 13 | 1595 | 70 | ||
AK162051.1 | - | 6 - 7 | 685 | 70 | ||
AK173147.1 | - | 11 - 12 | 1272 | 70 | ||
BC066788.1 | - | 12 - 13 | 1635 | 30049271 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
