Assay Details
Gene Symbol: Sap130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172965.2 | NP_766553.1 | 2 - 3 | 220 | 59 | |
GenBank mRNA | AK034401.1 | - | 2 - 3 | 224 | 59 | |
AK053675.1 | - | 2 - 3 | 221 | 59 | ||
AK084078.1 | - | 2 - 3 | 221 | 59 | ||
BC066030.1 | - | 2 - 3 | 232 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
