Assay Details
Gene Symbol: Sap130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172965.2 | NP_766553.1 | 5 - 6 | 735 | 91 | |
GenBank mRNA | AK034401.1 | - | 5 - 6 | 739 | 91 | |
AK053675.1 | - | 5 - 6 | 736 | 91 | ||
AK084078.1 | - | 5 - 6 | 736 | 91 | ||
BC063075.1 | - | 2 - 3 | 259 | 5721433 | 91 | |
BC066030.1 | - | 5 - 6 | 747 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
