Assay Details
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039866.1 | - | 2 - 3 | 279 | 95 | |
AK050894.1 | - | 5 - 6 | 721 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
