Assay Details
Gene Symbol: Ephb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173447.3 | NP_775623.3 | 10 - 11 | 2236 | 94 | |
GenBank mRNA | AK033966.1 | - | 10 - 11 | 2236 | 94 | |
AK036148.1 | - | 10 - 11 | 1924 | 94 | ||
AK135018.1 | - | 10 - 11 | 2202 | 94 | ||
BC057301.1 | - | 10 - 11 | 2176 | 6821724 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
