Assay Details
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174991.3 | NP_778156.2 | 5 - 6 | 2115 | 66 | |
GenBank mRNA | AK147456.1 | - | 5 - 6 | 2100 | 66 | |
AK147459.1 | - | 5 - 6 | 2068 | 66 | ||
AK147494.1 | - | 5 - 6 | 1902 | 66 | ||
AK147600.1 | - | 5 - 6 | 1877 | 66 | ||
AK147607.1 | - | 5 - 6 | 2119 | 66 | ||
AK220481.1 | - | 5 - 6 | 1494 | 66 | ||
AY168408.1 | - | 5 - 6 | 1627 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
