Assay Details
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174991.3 | NP_778156.2 | 19 - 20 | 3818 | 65 | |
GenBank mRNA | AK046831.1 | - | 3 - 4 | 336 | 65 | |
AK147456.1 | - | 19 - 20 | 3803 | 65 | ||
AK147459.1 | - | 19 - 20 | 3771 | 65 | ||
AK147494.1 | - | 19 - 20 | 3605 | 65 | ||
AK147600.1 | - | 19 - 20 | 3580 | 65 | ||
AK147607.1 | - | 19 - 20 | 3822 | 65 | ||
AK220481.1 | - | 19 - 20 | 3197 | 65 | ||
AY168408.1 | - | 19 - 20 | 3330 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
