Assay Details
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174991.3 | NP_778156.2 | 29 - 30 | 5338 | 61 | |
GenBank mRNA | AK046831.1 | - | 13 - 14 | 1856 | 61 | |
AK147456.1 | - | 29 - 30 | 5323 | 61 | ||
AK147459.1 | - | 29 - 30 | 5291 | 61 | ||
AK147494.1 | - | 29 - 30 | 5125 | 61 | ||
AK147600.1 | - | 28 - 29 | 5014 | 61 | ||
AK147607.1 | - | 29 - 30 | 5342 | 61 | ||
AK220481.1 | - | 29 - 30 | 4732 | 61 | ||
AY168408.1 | - | 29 - 30 | 4850 | 61 | ||
BC036963.1 | - | 11 - 12 | 1516 | 61 | ||
BC059857.1 | - | 8 - 9 | 1268 | 61 | ||
BC115835.1 | - | 5 - 6 | 1458 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
