Assay Details
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175628.3 | NP_783327.2 | 27 - 28 | 3459 | 75 | |
GenBank mRNA | AY185125.1 | - | 27 - 28 | 3405 | 75 | |
BC023844.1 | - | 17 - 18 | 2121 | 75 | ||
BC072642.1 | - | 27 - 28 | 3399 | 6839761 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
