Assay Details
Gene Symbol: Ap3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009680.3 | NP_033810.2 | 3 - 4 | 428 | 71 | |
GenBank mRNA | AF103809.2 | - | 3 - 4 | 365 | 71 | |
AK143559.1 | - | 3 - 4 | 424 | 71 | ||
AK163957.1 | - | 3 - 4 | 366 | 71 | ||
AK171595.1 | - | 3 - 4 | 429 | 71 | ||
BC015068.1 | - | 3 - 4 | 425 | 4018563 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
