Assay Details
Gene Symbol: Gfpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013528.3 | NP_038556.1 | 4 - 5 | 495 | 127 | |
GenBank mRNA | AF334736.1 | - | 4 - 5 | 452 | 127 | |
AK019852.1 | - | 4 - 5 | 491 | 127 | ||
BC002283.1 | - | 1 - 2 | 94 | 127 | ||
BC010516.1 | - | 4 - 5 | 443 | 4191553 | 127 | |
BC050762.1 | - | 4 - 5 | 491 | 6742987 | 127 | |
U00932.1 | - | 3 - 4 | 342 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
