Assay Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170537.1 | NP_001164008.1 | 1 - 2 | 345 | 87 | |
NM_025282.3 | NP_079558.1 | 1 - 2 | 345 | 87 | ||
GenBank mRNA | AK009139.1 | - | 1 - 2 | 185 | 87 | |
AK132573.1 | - | 1 - 2 | 174 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK009139.1, AK132573.1, NM_001170537.1, NM_025282.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
