Assay Details
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175168.4 | NP_780377.1 | 16 - 17 | 2803 | 71 | |
GenBank mRNA | AB055408.1 | - | 2 - 3 | 302 | 71 | |
AF531873.1 | - | 16 - 17 | 2653 | 71 | ||
AK053044.1 | - | 16 - 17 | 2736 | 71 | ||
AK163135.1 | - | 16 - 17 | 2736 | 71 | ||
BC027800.1 | - | 5 - 6 | 858 | 71 | ||
BC037175.1 | - | 1 - 2 | 445 | 71 | ||
BC076578.1 | - | 16 - 17 | 2803 | 30619280 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
