Assay Details
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175168.4 | NP_780377.1 | 1 - 2 | 239 | 67 | |
GenBank mRNA | AF531873.1 | - | 1 - 2 | 89 | 67 | |
AK053044.1 | - | 1 - 2 | 172 | 67 | ||
AK087891.1 | - | 1 - 2 | 173 | 67 | ||
AK163135.1 | - | 1 - 2 | 172 | 67 | ||
BC076578.1 | - | 1 - 2 | 239 | 30619280 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
