Assay Details
Gene Symbol: Catsperd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175350.3 | NP_780559.2 | 16 - 17 | 1549 | 78 | |
GenBank mRNA | AK077082.1 | - | 7 - 8 | 610 | 78 | |
BC115864.1 | - | 5 - 6 | 502 | 78 | ||
BC116633.1 | - | 6 - 7 | 616 | 40056655 | 78 | |
HQ441159.1 | - | 16 - 17 | 1541 | 78 | ||
HQ441160.1 | - | 13 - 14 | 1437 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
