Assay Details
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 12 - 13 | 1383 | 65 | |
GenBank mRNA | AK140362.1 | - | 12 - 13 | 1383 | 65 | |
BC038030.1 | - | 13 - 14 | 1394 | 4020725 | 65 | |
BC048005.1 | - | 8 - 9 | 738 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
