Assay Details
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177407.4 | NP_803126.1 | 7 - 8 | 666 | 112 | |
GenBank mRNA | AK083245.1 | - | 7 - 8 | 667 | 112 | |
AK122410.2 | - | 7 - 8 | 662 | 112 | ||
BC031745.1 | - | 7 - 8 | 647 | 4506005 | 112 | |
X14836.1 | - | 7 - 8 | 650 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
