Assay Details
Gene Symbol: Baiap2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177580.3 | NP_808248.1 | 10 - 11 | 1243 | 77 | |
GenBank mRNA | BC048937.1 | - | 10 - 11 | 1181 | 6396012 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
