Assay Details
Gene Symbol: Baiap2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177580.3 | NP_808248.1 | 2 - 3 | 249 | 83 | |
GenBank mRNA | AK143783.1 | - | 2 - 3 | 234 | 83 | |
BC048937.1 | - | 2 - 3 | 187 | 6396012 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
