Assay Details
Gene Symbol: Baiap2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177580.3 | NP_808248.1 | 6 - 7 | 586 | 62 | |
GenBank mRNA | AK143783.1 | - | 6 - 7 | 571 | 62 | |
BC048937.1 | - | 6 - 7 | 524 | 6396012 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
