Assay Details
Gene Symbol: Ccdc157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177616.3 | NP_808284.2 | 3 - 4 | 538 | 99 | |
GenBank mRNA | AK076934.1 | - | 3 - 4 | 466 | 99 | |
AK220409.1 | - | 3 - 4 | 519 | 99 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK076934.1, NM_177616.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
