Assay Details
Gene Symbol: Kansl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122738.1 | NP_001116210.1 | 10 - 11 | 2423 | 66 | |
NM_177645.4 | NP_808313.3 | 9 - 10 | 2396 | 66 | ||
GenBank mRNA | AK082897.1 | - | 9 - 10 | 1243 | 66 | |
AK220534.1 | - | 9 - 10 | 2347 | 66 | ||
BC141036.1 | - | 10 - 11 | 2306 | 9055648 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
